Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTERF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182501.3 | NP_872307.2 | 2 | 363 | ||
NR_028049.1 | 2 | 363 | ||||
XM_006712292.3 | XP_006712355.1 | 2 | 318 | |||
XM_006712294.1 | XP_006712357.1 | 2 | 318 | |||
XM_006712295.3 | XP_006712358.1 | 2 | 445 | |||
XM_011510634.2 | XP_011508936.1 | 2 | 620 | |||
XM_017003379.1 | XP_016858868.1 | 2 | 453 | |||
XM_017003380.1 | XP_016858869.1 | 2 | 453 | |||
XM_017003381.1 | XP_016858870.1 | 2 | 453 | |||
XM_017003382.1 | XP_016858871.1 | 2 | 453 | |||
XM_017003383.1 | XP_016858872.1 | 2 | 453 | |||
XM_017003384.1 | XP_016858873.1 | 2 | 453 | |||
XM_017003385.1 | XP_016858874.1 | 2 | 453 | |||
XM_017003386.1 | XP_016858875.1 | 2 | 924 | |||
XM_017003387.1 | XP_016858876.1 | 2 | 620 | |||
XM_017003388.1 | XP_016858877.1 | 2 | 620 | |||
XM_017003389.1 | XP_016858878.1 | 2 | 1398 | |||
XM_017003391.1 | XP_016858880.1 | 1 | 283 | |||
GenBank mRNA | AF161359.1 | - | 2 | 334 | ||
AK131298.1 | - | 2 | 1137 | |||
AK291665.1 | - | 2 | 318 | |||
AK296515.1 | - | 2 | 341 | |||
BC053533.1 | - | 2 | 316 | 4417008 | ||
BX648184.1 | - | 2 | 329 | |||
CD388157.1 | - | 2 | 353 |