Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTERF4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182501.3 | NP_872307.2 | 4 | 845 | ||
NR_028049.1 | 4 | 845 | ||||
XM_011510634.2 | XP_011508936.1 | 4 | 1102 | |||
XM_017003379.1 | XP_016858868.1 | 5 | 1079 | |||
XM_017003380.1 | XP_016858869.1 | 5 | 1079 | |||
XM_017003381.1 | XP_016858870.1 | 5 | 1079 | |||
XM_017003382.1 | XP_016858871.1 | 5 | 1079 | |||
XM_017003383.1 | XP_016858872.1 | 5 | 1079 | |||
XM_017003384.1 | XP_016858873.1 | 5 | 1079 | |||
XM_017003385.1 | XP_016858874.1 | 5 | 1079 | |||
XM_017003386.1 | XP_016858875.1 | 5 | 1550 | |||
XM_017003387.1 | XP_016858876.1 | 4 | 1102 | |||
XM_017003388.1 | XP_016858877.1 | 4 | 1102 | |||
XM_017003390.1 | XP_016858879.1 | 3 | 335 | |||
XM_017003391.1 | XP_016858880.1 | 3 | 1355 | |||
GenBank mRNA | AK131298.1 | - | 4 | 1619 | ||
AK291665.1 | - | 4 | 800 | |||
AK294192.1 | - | 3 | 301 | |||
BC053533.1 | - | 4 | 798 | 4417008 |