Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYZL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304386.1 | NP_001291315.1 | 2, 3 | 390 | ||
NM_144634.3 | NP_653235.1 | 2, 3 | 409 | |||
XM_011533355.2 | XP_011531657.1 | 2, 3 | 452 | |||
XM_017005705.1 | XP_016861194.1 | 4, 5 | 764 | |||
XM_017005706.1 | XP_016861195.1 | 3, 4 | 522 | |||
GenBank mRNA | AF326749.1 | - | 2, 3 | 338 | ||
BC016747.2 | - | 2, 3 | 380 | 4830396 | ||
BX282166.1 | - | 2, 3 | 342 |