Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DCBLD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080927.3 | NP_563615.3 | 12 | 1901 | ||
XM_011512419.2 | XP_011510721.1 | 11 | 1617 | |||
GenBank mRNA | AB073146.1 | - | 12 | 1901 | ||
AF387547.1 | - | 12 | 1549 | |||
AK056350.1 | - | 12 | 1894 | |||
AK302649.1 | - | 12 | 1381 | |||
BC029658.1 | - | 12 | 1420 | 4431275 | ||
BX647140.1 | - | 3 | 4134 |