Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM43A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153690.4 | NP_710157.2 | 1 | 955 | ||
GenBank mRNA | AF289585.1 | - | 1 | 609 | ||
AK074503.1 | - | 1 | 121 | |||
BC039689.2 | - | 1 | 164 | 4906944 | ||
BC064989.1 | - | 1 | 556 | 5423287 |