Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UROC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165974.1 | NP_001159446.1 | 15 | 1654 | ||
NM_144639.2 | NP_653240.1 | 14 | 1474 | |||
GenBank mRNA | AB075869.1 | - | 15 | 1634 | ||
AK055862.1 | - | 14 | 1452 | |||
BC115405.1 | - | 14 | 1452 | 40076084 | ||
BC115406.1 | - | 14 | 1452 | 40076086 |