Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLYCTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144951.1 | NP_001138423.1 | 4 | 733 | ||
NM_145262.3 | NP_660305.2 | 5 | 909 | |||
NR_026699.1 | 5 | 911 | ||||
NR_026700.1 | 4 | 493 | ||||
NR_026701.1 | 5 | 909 | ||||
XM_017005730.1 | XP_016861219.1 | 4 | 755 | |||
GenBank mRNA | AF448855.1 | - | 5 | 910 | ||
AY134474.1 | - | 3 | 615 | |||
AY295075.1 | - | 3 | 666 | |||
BC021896.1 | - | 4 | 671 | 3917295 | ||
BC036862.1 | - | 5 | 911 | |||
BC042151.1 | - | 5 | 873 | 4577225 | ||
DQ352863.1 | - | 4 | 813 |