Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SYNPR | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130003.1 | NP_001123475.1 | 6 | 1138 | ||
NM_144642.4 | NP_653243.1 | 5 | 1151 | |||
XM_017005731.1 | XP_016861220.1 | 6 | 904 | |||
GenBank mRNA | AF411860.2 | - | 5 | 900 | ||
AK122831.1 | - | 6 | 928 | |||
AK293628.1 | - | 5 | 850 | |||
AK309017.1 | - | 5 | 933 | |||
AK312465.1 | - | 5 | 1056 | |||
AK315958.1 | - | 4 | 915 | |||
AL834457.1 | - | 5 | 880 | |||
BC022518.1 | - | 5 | 900 | 4792167 | ||
CR936762.1 | - | 6 | 1059 |