Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SCLT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300897.1 | NP_001287826.1 | 4 | 716 | ||
NM_001300898.1 | NP_001287827.1 | 4 | 716 | |||
NM_144643.3 | NP_653244.2 | 4 | 716 | |||
XM_017007716.1 | XP_016863205.1 | 4 | 716 | |||
XM_017007717.1 | XP_016863206.1 | 4 | 716 | |||
XM_017007718.1 | XP_016863207.1 | 4 | 716 | |||
XM_017007719.1 | XP_016863208.1 | 4 | 237 | |||
XM_017007720.1 | XP_016863209.1 | 4 | 583 | |||
GenBank mRNA | AK055217.1 | - | 4 | 716 | ||
AL557810.3 | - | 4 | 597 | |||
BC064428.1 | - | 4 | 654 | |||
BC121057.1 | - | 4 | 522 | |||
BC121058.1 | - | 4 | 519 | |||
BC128051.1 | - | 4 | 521 |