Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZFP42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304358.1 | NP_001291287.1 | 3 | 351 | ||
NM_174900.4 | NP_777560.2 | 4 | 517 | |||
XM_011531604.2 | XP_011529906.1 | 3 | 372 | |||
XM_011531605.1 | XP_011529907.1 | 4 | 522 | |||
XM_011531606.1 | XP_011529908.1 | 4 | 526 | |||
XM_011531607.2 | XP_011529909.1 | 3 | 369 | |||
GenBank mRNA | AF450454.1 | - | 1 | 109 | ||
AK056719.1 | - | 3 | 351 | |||
AY429593.1 | - | 4 | 517 |