Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPATA18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297608.1 | NP_001284537.1 | 7 | 1396 | ||
NM_145263.3 | NP_660306.1 | 8 | 1492 | |||
NR_123728.1 | 8 | 1492 | ||||
XM_005265716.1 | XP_005265773.1 | 6 | 1198 | |||
XM_005265718.1 | XP_005265775.1 | 4 | 980 | |||
XM_011534369.1 | XP_011532671.1 | 7 | 1294 | |||
XM_011534370.1 | XP_011532672.1 | 8 | 1418 | |||
XM_011534372.1 | XP_011532674.1 | 5 | 1072 | |||
XM_017007731.1 | XP_016863220.1 | 6 | 890 | |||
GenBank mRNA | AB465501.1 | - | 8 | 1118 | ||
AB465502.1 | - | 7 | 1022 | |||
AK057468.1 | - | 8 | 1404 | |||
AK098625.1 | - | 5 | 717 | |||
AK304386.1 | - | 7 | 1311 | |||
AK314799.1 | - | 8 | 1362 | |||
BC025396.1 | - | 8 | 1399 | 4821894 | ||
BC037886.1 | - | 2 | 171 |