Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AASDH | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286668.1 | NP_001273597.1 | 14 | 3009 | ||
NM_001286669.1 | NP_001273598.1 | 14 | 3160 | |||
NM_001286670.1 | NP_001273599.1 | 12 | 2684 | |||
NM_001286671.1 | NP_001273600.1 | 12 | 2862 | |||
NM_001323890.1 | NP_001310819.1 | 16 | 3398 | |||
NM_001323892.1 | NP_001310821.1 | 14 | 3117 | |||
NM_001323893.1 | NP_001310822.1 | 13 | 2996 | |||
NM_001323899.1 | NP_001310828.1 | 14 | 3174 | |||
NM_181806.3 | NP_861522.2 | 15 | 3281 | |||
XM_017007740.1 | XP_016863229.1 | 14 | 3110 | |||
XM_017007741.1 | XP_016863230.1 | 16 | 3357 | |||
XM_017007742.1 | XP_016863231.1 | 13 | 3057 | |||
XM_017007743.1 | XP_016863232.1 | 13 | 2946 | |||
XM_017007744.1 | XP_016863233.1 | 12 | 2864 | |||
GenBank mRNA | AA743758.1 | - | 1 | 330 | ||
AF516672.1 | - | 14 | 2924 | |||
AK304309.1 | - | 15 | 3272 | |||
AK316296.1 | - | 15 | 3262 | |||
AY422212.1 | - | 15 | 3196 | |||
BC142709.1 | - | 12 | 2798 | 40148820 | ||
BX640635.1 | - | 10 | 4112 |