Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IL31RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242636.1 | NP_001229565.1 | 7 | 1016 | ||
NM_001242637.1 | NP_001229566.1 | 7 | 1025 | |||
NM_001242638.1 | NP_001229567.1 | 9 | 1389 | |||
NM_001242639.1 | NP_001229568.1 | 8 | 1160 | |||
NM_001297570.1 | NP_001284499.1 | 7 | 1025 | |||
NM_001297572.1 | NP_001284501.1 | 7 | 1153 | |||
NM_139017.5 | NP_620586.3 | 7 | 1025 | |||
XM_011543142.2 | XP_011541444.1 | 7 | 858 | |||
XM_011543143.2 | XP_011541445.1 | 7 | 1048 | |||
XM_011543144.2 | XP_011541446.1 | 6 | 930 | |||
XM_011543145.2 | XP_011541447.1 | 4 | 499 | |||
GenBank mRNA | AF106913.1 | - | 7 | 889 | ||
AF486620.1 | - | 6 | 737 | |||
AK297938.1 | - | 7 | 1137 | |||
AY358117.1 | - | 6 | 737 | |||
AY358740.1 | - | 7 | 1025 | |||
AY499339.1 | - | 7 | 898 | |||
AY499340.1 | - | 7 | 898 | |||
AY499341.1 | - | 7 | 898 | |||
AY499342.1 | - | 9 | 1272 | |||
BC110490.2 | - | 8 | 952 | 40007489 |