Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM173B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258389.1 | NP_001245318.1 | 4 | 456 | ||
NM_199133.3 | NP_954584.2 | 4 | 500 | |||
NR_047669.1 | 5 | 703 | ||||
NR_047670.1 | 5 | 592 | ||||
XM_011513964.1 | XP_011512266.1 | 4 | 487 | |||
GenBank mRNA | AK055604.1 | - | 5 | 580 | ||
AK300042.1 | - | 4 | 477 | |||
AK316317.1 | - | 4 | 485 | |||
BC063406.1 | - | 4 | 468 | 5190223 | ||
BG611454.1 | - | 5 | 684 | |||
DA523490.1 | - | 4 | 435 |