Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DNAJC21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012339.2 | NP_001012339.2 | 7, 8 | 1219 | ||
NM_194283.3 | NP_919259.3 | 7, 8 | 1219 | |||
XM_005248249.4 | XP_005248306.2 | 7, 8 | 1272 | |||
XM_005248250.3 | XP_005248307.2 | 7, 8 | 1272 | |||
XM_011513965.2 | XP_011512267.2 | 7, 8 | 1272 | |||
XM_011513966.2 | XP_011512268.2 | 7, 8 | 1272 | |||
GenBank mRNA | AK127749.1 | - | 4, 5 | 649 | ||
AY177688.1 | - | 7, 8 | 1228 | |||
AY258422.1 | - | 7, 8 | 1222 | |||
BC050472.1 | - | 3, 4 | 519 | |||
BC107577.1 | - | 4, 5 | 619 |