Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POC5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099271.1 | NP_001092741.1 | 7, 8 | 999 | ||
NM_152408.2 | NP_689621.2 | 6, 7 | 798 | |||
XM_005248436.1 | XP_005248493.1 | 7, 8 | 999 | |||
XM_011543158.2 | XP_011541460.1 | 7, 8 | 999 | |||
XM_011543159.1 | XP_011541461.1 | 8, 9 | 1005 | |||
XM_011543160.1 | XP_011541462.1 | 6, 7 | 859 | |||
XM_017009038.1 | XP_016864527.1 | 6, 7 | 919 | |||
XM_017009039.1 | XP_016864528.1 | 6, 7 | 838 | |||
GenBank mRNA | AK093098.1 | - | 6, 7 | 768 | ||
AK296071.1 | - | 7, 8 | 916 | |||
BC101325.1 | - | 6, 7 | 764 | 40024984 | ||
BC101326.2 | - | 6, 7 | 768 | 40024987 | ||
BC101327.2 | - | 6, 7 | 768 | 40024989 | ||
BC101328.2 | - | 6, 7 | 765 |