Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127715.2 | NP_001121187.1 | 16 | 1913 | ||
NM_139244.4 | NP_640337.3 | 16 | 1913 | |||
XM_006715339.2 | XP_006715402.1 | 16 | 2087 | |||
XM_011535453.1 | XP_011533755.1 | 16 | 1685 | |||
XM_017010267.1 | XP_016865756.1 | 11 | 1984 | |||
XM_017010268.1 | XP_016865757.1 | 16 | 1950 | |||
GenBank mRNA | AB073398.1 | - | 7 | 752 | ||
AK090549.1 | - | 6 | 623 | |||
AK128121.1 | - | 16 | 1680 | |||
AL834152.1 | - | 16 | 1899 | |||
BC113382.1 | - | 16 | 1786 | 8322434 | ||
BC113408.1 | - | 16 | 1786 | 8322460 |