Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STXBP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001127715.2 | NP_001121187.1 | 23 | 2571 | ||
NM_139244.4 | NP_640337.3 | 21 | 2463 | |||
XM_006715339.2 | XP_006715402.1 | 22 | 2697 | |||
XM_011535453.1 | XP_011533755.1 | 22 | 2295 | |||
XM_017010267.1 | XP_016865756.1 | 18 | 2642 | |||
GenBank mRNA | AB073398.1 | - | 12 | 1302 | ||
AK055484.1 | - | 4 | 281 | |||
AK090549.1 | - | 13 | 1281 | |||
AK128121.1 | - | 22 | 2290 | |||
AL834152.1 | - | 24 | 2620 | |||
BC113382.1 | - | 21 | 2336 | 8322434 | ||
BC113408.1 | - | 21 | 2336 | 8322460 |