Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD109 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159587.2 | NP_001153059.1 | 27 | 3558 | ||
NM_001159588.2 | NP_001153060.1 | 26 | 3327 | |||
NM_133493.4 | NP_598000.2 | 27 | 3558 | |||
XM_005248659.3 | XP_005248716.1 | 28 | 3705 | |||
XM_011535473.1 | XP_011533775.1 | 27 | 3403 | |||
XM_017010283.1 | XP_016865772.1 | 25 | 3170 | |||
GenBank mRNA | AF410459.1 | - | 27 | 3539 | ||
AK123960.1 | - | 13 | 1636 | |||
AK313636.1 | - | 27 | 3538 | |||
AL834478.1 | - | 13 | 1654 | |||
AY149920.1 | - | 27 | 3427 | |||
AY788891.1 | - | 27 | 3452 | |||
BX641095.1 | - | 26 | 3214 | |||
EF553520.1 | - | 27 | 3457 |