Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C6orf141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145652.1 | NP_001139124.1 | 1 | 1106 | ||
XM_005248850.3 | XP_005248907.1 | 1 | 862 | |||
XM_005248852.2 | XP_005248909.1 | 1 | 862 | |||
XM_005248853.2 | XP_005248910.1 | 1 | 862 | |||
XM_006714997.2 | XP_006715060.1 | 1 | 862 | |||
XM_011514303.2 | XP_011512605.1 | 1 | 862 | |||
XM_017010294.1 | XP_016865783.1 | 1 | 862 | |||
XM_017010295.1 | XP_016865784.1 | 1 | 862 | |||
XM_017010296.1 | XP_016865785.1 | 1 | 862 | |||
XM_017010297.1 | XP_016865786.1 | 1 | 862 | |||
XM_017010298.1 | XP_016865787.1 | 1 | 862 | |||
GenBank mRNA | AK054918.1 | - | 1 | 831 | ||
AK289889.1 | - | 1 | 860 | |||
BC036917.1 | - | 1 | 840 | 5201433 |