Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TRIM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001281450.1 | NP_001268379.1 | 2 | 585 | ||
NM_001281451.1 | NP_001268380.1 | 2 | 513 | |||
NM_178125.3 | NP_835226.2 | 2 | 585 | |||
XM_011515787.1 | XP_011514089.1 | 2 | 513 | |||
XM_011515789.2 | XP_011514091.2 | 2 | 513 | |||
GenBank mRNA | AK057366.1 | - | 2 | 585 | ||
AK292074.1 | - | 2 | 511 | |||
AY081948.1 | - | 2 | 585 | |||
AY081949.1 | - | 2 | 585 | |||
BC112152.1 | - | 2 | 427 | 8327620 | ||
BC112154.1 | - | 2 | 427 | 8327622 | ||
BC143787.1 | - | 2 | 427 | 9052301 |