Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM139 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242773.2 | NP_001229702.1 | 3 | 336 | ||
NM_001242774.2 | NP_001229703.1 | 4 | 447 | |||
NM_001242775.2 | NP_001229704.1 | 3 | 360 | |||
NM_001282876.1 | NP_001269805.1 | 2 | 658 | |||
NM_001282877.1 | NP_001269806.1 | 3 | 339 | |||
NM_153345.3 | NP_699176.1 | 2 | 238 | |||
GenBank mRNA | AK027352.1 | - | 1 | 1148 | ||
AK075067.1 | - | 2 | 183 | |||
AK172816.1 | - | 4 | 389 | |||
AK315170.1 | - | 3 | 284 | |||
AY358781.1 | - | 2 | 253 | |||
BC035517.1 | - | 2 | 638 | 5207904 | ||
DA626625.1 | - | 3 | 299 | |||
HY180523.1 | - | 3 | 340 |
Gene Symbol: LOC105375545 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_133932.1 | 1 | 229 | |||
GenBank mRNA | DA339204.1 | - | 1 | 203 | ||
DB173620.1 | - | 1 | 231 |