Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM68 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286657.1 | NP_001273586.1 | 4 | 716 | ||
NM_001286661.1 | NP_001273590.1 | 3 | 416 | |||
NM_152417.2 | NP_689630.1 | 5 | 855 | |||
XM_005251150.1 | XP_005251207.1 | 5 | 844 | |||
XM_011517454.1 | XP_011515756.1 | 5 | 868 | |||
XM_011517455.2 | XP_011515757.1 | 5 | 844 | |||
XM_017013040.1 | XP_016868529.1 | 4 | 428 | |||
GenBank mRNA | AK056932.1 | - | 5 | 774 | ||
AL832935.1 | - | 3 | 325 | |||
CN272366.1 | - | 3 | 343 |