Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PXDNL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144651.4 | NP_653252.3 | 17 | 2785 | ||
XM_005251168.3 | XP_005251225.1 | 3 | 932 | |||
XM_006716420.3 | XP_006716483.1 | 15 | 4083 | |||
XM_011517456.2 | XP_011515758.1 | 16 | 4299 | |||
XM_011517457.2 | XP_011515759.1 | 16 | 4299 | |||
XM_011517458.2 | XP_011515760.1 | 15 | 2459 | |||
XM_017013041.1 | XP_016868530.1 | 14 | 2420 | |||
GenBank mRNA | AK131524.1 | - | 1 | 482 | ||
AY877349.1 | - | 17 | 2785 | |||
BC132813.1 | - | 1 | 305 | |||
EU170240.1 | - | 17 | 2684 | |||
KJ545488.1 | - | 15 | 2496 |