Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LETM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199659.2 | NP_001186588.1 | 8 | 1258 | ||
NM_001199660.2 | NP_001186589.1 | 8 | 1097 | |||
NM_001286787.1 | NP_001273716.1 | 8 | 1244 | |||
NM_001286819.1 | NP_001273748.1 | 8 | 1329 | |||
NM_144652.3 | NP_653253.1 | 7 | 1114 | |||
XM_006716291.3 | XP_006716354.1 | 8 | 1219 | |||
XM_011544406.1 | XP_011542708.1 | 8 | 1218 | |||
XM_011544407.1 | XP_011542709.1 | 8 | 1180 | |||
XM_011544408.2 | XP_011542710.1 | 8 | 3825 | |||
XM_011544409.2 | XP_011542711.1 | 8 | 1498 | |||
XM_011544410.2 | XP_011542712.1 | 5 | 746 | |||
XM_017013051.1 | XP_016868540.1 | 8 | 1561 | |||
XM_017013052.1 | XP_016868541.1 | 8 | 1289 | |||
XM_017013053.1 | XP_016868542.1 | 8 | 3635 | |||
XM_017013054.1 | XP_016868543.1 | 8 | 1274 | |||
XM_017013055.1 | XP_016868544.1 | 8 | 1417 | |||
XM_017013056.1 | XP_016868545.1 | 8 | 1263 | |||
XM_017013057.1 | XP_016868546.1 | 7 | 3679 | |||
GenBank mRNA | AK058138.1 | - | 7 | 1053 | ||
AK302317.1 | - | 8 | 1197 | |||
AK302474.1 | - | 8 | 1069 | |||
AY173945.1 | - | 8 | 1189 | |||
BC029541.1 | - | 5 | 682 | 5297908 | ||
BC069830.1 | - | 8 | 1178 | |||
CB242043.1 | - | 4 | 593 |