Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CARNMT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320497.1 | NP_001307426.1 | 7 | 946 | ||
NM_152420.2 | NP_689633.1 | 7 | 1266 | |||
NR_135282.1 | 6 | 1125 | ||||
XM_005251701.3 | XP_005251758.2 | 7 | 941 | |||
XM_017014269.1 | XP_016869758.1 | 7 | 1253 | |||
XM_017014270.1 | XP_016869759.1 | 8 | 1199 | |||
XM_017014271.1 | XP_016869760.1 | 8 | 1691 | |||
GenBank mRNA | AK098661.1 | - | 6 | 693 | ||
AK299691.1 | - | 6 | 968 | |||
BC034033.1 | - | 7 | 1230 | 4278547 | ||
BX538061.1 | - | 6 | 783 | |||
BX647386.1 | - | 7 | 1035 |