Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024708.3 | NP_078984.2 | 4, 5 | 983 | ||
NM_198243.2 | NP_937886.1 | 4, 5 | 983 | |||
GenBank mRNA | AF451994.1 | - | 4, 5 | 903 | ||
AK026204.1 | - | 4, 5 | 903 | |||
AK222958.1 | - | 4, 5 | 903 | |||
AK222974.1 | - | 4, 5 | 903 | |||
AK289860.1 | - | 4, 5 | 936 | |||
BC063581.1 | - | 4, 5 | 924 | 4370796 |