Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: S100A16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001317007.1 | NP_001303936.1 | 3 | 696 | ||
NM_001317008.1 | NP_001303937.1 | 3 | 417 | |||
NM_080388.2 | NP_525127.1 | 3 | 456 | |||
NR_133645.1 | 3 | 482 | ||||
GenBank mRNA | AK290804.1 | - | 3 | 346 | ||
AW954845.1 | - | 3 | 458 | |||
AY221961.1 | - | 3 | 327 | |||
AY762098.1 | - | 3 | 371 | |||
BC010541.1 | - | 3 | 326 | 3458906 | ||
BC019099.1 | - | 3 | 370 | 5013080 | ||
BC095462.1 | - | 3 | 291 | 30553208 | ||
BM739127.1 | - | 3 | 458 | |||
HY178745.1 | - | 3 | 417 | |||
U92985.1 | - | 3 | 307 |