Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NEK7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133494.2 | NP_598001.1 | 9 | 1103 | ||
XM_017000344.1 | XP_016855833.1 | 9 | 1105 | |||
XM_017000345.1 | XP_016855834.1 | 10 | 1212 | |||
GenBank mRNA | AB062450.1 | - | 8 | 822 | ||
AK313411.1 | - | 9 | 903 |