Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C20orf96 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080571.1 | NP_542138.1 | 5 | 576 | ||
NM_153269.2 | NP_695001.2 | 5 | 572 | |||
XM_006723543.3 | XP_006723606.1 | 4 | 502 | |||
GenBank mRNA | AK091924.1 | - | 3 | 382 | ||
AK097394.1 | - | 5 | 543 | |||
AK302223.1 | - | 3 | 290 | |||
AK304526.1 | - | 3 | 397 | |||
BC051762.1 | - | 4 | 521 | |||
BC134417.1 | - | 5 | 530 | 40115967 | ||
BC137450.1 | - | 5 | 539 | 9021450 | ||
BC144648.1 | - | 4 | 490 | |||
BC144649.1 | - | 5 | 543 | 9053180 | ||
BX640715.1 | - | 4 | 516 | |||
EL734555.1 | - | 3 | 245 |