Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOL4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256798.1 | NP_001243727.1 | 10 | 1990 | ||
NM_080616.4 | NP_542183.2 | 7 | 1329 | |||
XM_005260282.4 | XP_005260339.1 | 10 | 2276 | |||
XM_005260283.4 | XP_005260340.1 | 11 | 2300 | |||
XM_005260284.4 | XP_005260341.1 | 10 | 1575 | |||
XM_005260285.4 | XP_005260342.2 | 10 | 2280 | |||
XM_005260286.4 | XP_005260343.1 | 10 | 2200 | |||
XM_005260288.2 | XP_005260345.1 | 7 | 1377 | |||
XM_005260289.4 | XP_005260346.1 | 7 | 1328 | |||
XM_006723702.3 | XP_006723765.1 | 11 | 1667 | |||
XM_006723703.3 | XP_006723766.1 | 10 | 1579 | |||
XM_011528562.2 | XP_011526864.1 | 10 | 1984 | |||
XM_011528563.1 | XP_011526865.1 | 10 | 1984 | |||
XM_017027669.1 | XP_016883158.1 | 12 | 2321 | |||
XM_017027670.1 | XP_016883159.1 | 7 | 1232 | |||
GenBank mRNA | AK056286.1 | - | 7 | 1132 | ||
BC065370.1 | - | 6 | 1101 | 6261963 |