Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBE2F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278305.1 | NP_001265234.1 | 9, 10 | 643 | ||
NM_001278306.1 | NP_001265235.1 | 8, 9 | 580 | |||
NM_001278307.1 | NP_001265236.1 | 7, 8 | 618 | |||
NM_080678.2 | NP_542409.1 | 9, 10 | 714 | |||
NR_103498.1 | 8, 9 | 646 | ||||
NR_103499.1 | 7, 8 | 623 | ||||
NR_103500.1 | 10, 11 | 813 | ||||
GenBank mRNA | AF310723.1 | - | 8, 9 | 510 | ||
AK290063.1 | - | 9, 10 | 625 | |||
AK294107.1 | - | 8, 9 | 526 | |||
AK297502.1 | - | 7, 8 | 549 | |||
AK303094.1 | - | 7, 8 | 572 | |||
BC010549.1 | - | 9, 10 | 601 | 4153270 | ||
BC039435.1 | - | 1, 2 | 813 | |||
BM470609.1 | - | 444 |