Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NANP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152667.2 | NP_689880.1 | 2 | 666 | ||
GenBank mRNA | AK055472.1 | - | 2 | 633 | ||
AL833116.1 | - | 2 | 640 | |||
BC022552.1 | - | 2 | 632 | 4819016 |