Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIRPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040022.1 | NP_001035111.1 | 9 | 1714 | ||
NM_001040023.1 | NP_001035112.1 | 8 | 1622 | |||
NM_080792.2 | NP_542970.1 | 9 | 1381 | |||
XM_005260669.3 | XP_005260726.1 | 9 | 1776 | |||
XM_005260670.3 | XP_005260727.1 | 9 | 1392 | |||
XM_006723545.3 | XP_006723608.1 | 6 | 963 | |||
XM_011529173.2 | XP_011527475.1 | 8 | 1381 | |||
GenBank mRNA | AB023430.1 | - | 9 | 1381 | ||
AK290776.1 | - | 9 | 1487 | |||
AK298243.1 | - | 9 | 1332 | |||
AK312521.1 | - | 8 | 1435 | |||
BC026692.1 | - | 9 | 1398 | 3916836 | ||
BC033092.1 | - | 8 | 1386 | 4589298 | ||
BC038510.2 | - | 8 | 1619 | 5261789 | ||
BC075849.1 | - | 9 | 1560 | 4589732 | ||
D86043.1 | - | 8 | 1353 | |||
Y10375.1 | - | 8 | 1363 | |||
Y11047.1 | - | 7 | 1129 |