Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SREK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077199.2 | NP_001070667.1 | 11 | 1870 | ||
NM_001270492.1 | NP_001257421.1 | 12 | 1991 | |||
NM_001323527.1 | NP_001310456.1 | 12 | 1994 | |||
NM_001323529.1 | NP_001310458.1 | 11 | 1867 | |||
NM_001323534.1 | NP_001310463.1 | 12 | 1885 | |||
NM_001323535.1 | NP_001310464.1 | 11 | 1758 | |||
NM_139168.3 | NP_631907.1 | 12 | 2023 | |||
XM_011543171.2 | XP_011541473.1 | 11 | 2095 | |||
XM_017009055.1 | XP_016864544.1 | 10 | 1404 | |||
XM_017009056.1 | XP_016864545.1 | 11 | 1655 | |||
GenBank mRNA | AF459094.1 | - | 11 | 1704 | ||
AK091758.1 | - | 11 | 1752 | |||
AK125893.1 | - | 4 | 1555 | |||
AK308287.1 | - | 12 | 1893 | |||
BC067770.1 | - | 12 | 2024 | 4827589 | ||
BC112343.1 | - | 9 | 1392 | 40008558 |