Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASB14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142733.2 | NP_001136205.2 | 8 | 1331 | ||
NM_130387.5 | NP_569058.1 | 1 | 355 | |||
XM_017005736.1 | XP_016861225.1 | 8 | 1681 | |||
XM_017005737.1 | XP_016861226.1 | 8 | 1645 | |||
GenBank mRNA | AF403032.1 | - | 1 | 355 | ||
AL832472.1 | - | 8 | 1332 |