Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDLRAD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304263.1 | NP_001291192.1 | 2 | 254 | ||
NM_001304264.1 | NP_001291193.1 | 2 | 254 | |||
NM_174902.3 | NP_777562.1 | 3 | 401 | |||
GenBank mRNA | AK075546.1 | - | 3 | 320 | ||
AK293930.1 | - | 2 | 196 | |||
AK299186.1 | - | 3 | 320 | |||
BC042754.1 | - | 3 | 304 | 4828259 | ||
BC136286.1 | - | 3 | 401 | 9020273 | ||
BC143823.1 | - | 2 | 254 | |||
BC143824.1 | - | 2 | 254 | 9052339 |