Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CWF19L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152434.2 | NP_689647.2 | 8 | 1151 | ||
XM_011542620.2 | XP_011540922.1 | 8 | 1149 | |||
GenBank mRNA | AK056905.1 | - | 6 | 803 | ||
AK126881.1 | - | 8 | 1144 | |||
AK293001.1 | - | 8 | 1086 | |||
BC070142.1 | - | 8 | 1130 | |||
BC107418.1 | - | 8 | 1134 | |||
BC110439.1 | - | 5 | 764 | 40001346 | ||
BC110440.1 | - | 5 | 764 | 40001348 | ||
BC110441.1 | - | 5 | 764 | 40001349 | ||
BC110442.1 | - | 5 | 764 | 40001350 | ||
BC118669.1 | - | 5 | 767 | 40012545 | ||
BX647832.1 | - | 8 | 1125 | |||
BX648704.1 | - | 8 | 1130 | |||
BX649022.1 | - | 8 | 1121 |