Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138471.2 | NP_612480.1 | 6 | 1397 | ||
XM_006718437.1 | XP_006718500.1 | 7 | 1455 | |||
XM_011544770.1 | XP_011543072.1 | 7 | 1417 | |||
XM_011544771.1 | XP_011543073.1 | 6 | 1291 | |||
XM_011544772.1 | XP_011543074.1 | 6 | 1359 | |||
GenBank mRNA | AK091493.1 | - | 1 | 2796 | ||
AK092098.1 | - | 1 | 1442 | |||
BC002782.2 | - | 6 | 1155 | 3621749 | ||
BC007540.1 | - | 6 | 1155 | 3609337 | ||
BC056402.1 | - | 6 | 1320 | 5764461 | ||
CR749697.1 | - | 2 | 216 |