Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf84 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138471.2 | NP_612480.1 | 1 | 354 | ||
XM_005273782.2 | XP_005273839.1 | 1 | 354 | |||
XM_005273783.2 | XP_005273840.1 | 1 | 354 | |||
XM_006718437.1 | XP_006718500.1 | 1 | 354 | |||
XM_011544770.1 | XP_011543072.1 | 1 | 354 | |||
XM_011544771.1 | XP_011543073.1 | 1 | 354 | |||
XM_011544772.1 | XP_011543074.1 | 1 | 354 | |||
GenBank mRNA | BC002782.2 | - | 1 | 112 | 3621749 | |
BC007540.1 | - | 1 | 112 | 3609337 | ||
BC056402.1 | - | 1 | 277 | 5764461 | ||
HY013538.1 | - | 1 | 355 |