Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLEKHA7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_175058.4 | NP_778228.3 | 9 | 870 | ||
XM_017017240.1 | XP_016872729.1 | 9 | 877 | |||
XM_017017241.1 | XP_016872730.1 | 9 | 877 | |||
XM_017017242.1 | XP_016872731.1 | 9 | 877 | |||
XM_017017243.1 | XP_016872732.1 | 9 | 877 | |||
GenBank mRNA | BC071599.1 | - | 9 | 866 | 30343461 |