Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GLIPR1L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270396.1 | NP_001257325.1 | 2 | 502 | ||
NM_152436.2 | NP_689649.1 | 2 | 502 | |||
NR_072995.1 | 3 | 632 | ||||
XM_011537949.2 | XP_011536251.1 | 2 | 416 | |||
XM_011537950.2 | XP_011536252.1 | 2 | 388 | |||
XM_017018848.1 | XP_016874337.1 | 3 | 492 | |||
XM_017018849.1 | XP_016874338.1 | 3 | 518 | |||
GenBank mRNA | AK093175.1 | - | 2 | 492 | ||
BC016749.1 | - | 2 | 509 | |||
BC029557.1 | - | 2 | 463 | 5262538 | ||
BX640916.1 | - | 3 | 601 |