Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYRM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001660.2 | NP_001001660.2 | 3 | 343 | ||
XM_005253319.4 | XP_005253376.1 | 3 | 346 | |||
XM_005253320.4 | XP_005253377.1 | 4 | 450 | |||
XM_017018850.1 | XP_016874339.1 | 3 | 1764 | |||
GenBank mRNA | AK057730.1 | - | 3 | 347 | ||
AK095164.1 | - | 2 | 1844 | |||
BC071769.1 | - | 3 | 277 | 4246777 | ||
BC100276.1 | - | 3 | 294 | 6178563 | ||
BG771497.1 | - | 3 | 349 |