Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CPNE8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153634.2 | NP_705898.1 | 19 | 1544 | ||
XM_017018851.1 | XP_016874340.1 | 20 | 2517 | |||
XM_017018852.1 | XP_016874341.1 | 19 | 1490 | |||
GenBank mRNA | AK074223.1 | - | 5 | 406 | ||
AK094998.1 | - | 7 | 1118 | |||
AK098593.1 | - | 19 | 1475 | |||
AK307908.1 | - | 19 | 1515 | |||
AY177785.1 | - | 19 | 1460 | |||
BC046366.1 | - | 7 | 861 | |||
BC110577.1 | - | 19 | 1535 | 40007743 | ||
BC110578.1 | - | 19 | 1535 | 40007744 | ||
BX538055.1 | - | 19 | 1558 |