Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAD9B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286531.1 | NP_001273460.1 | 8, 9 | 887 | ||
NM_001286532.1 | NP_001273461.1 | 8, 9 | 909 | |||
NM_001286533.1 | NP_001273462.1 | 9, 10 | 1065 | |||
NM_001286534.1 | NP_001273463.1 | 9, 10 | 985 | |||
NM_001286535.1 | NP_001273464.1 | 8, 9 | 916 | |||
NM_001286536.1 | NP_001273465.1 | 6, 7 | 709 | |||
NM_152442.3 | NP_689655.3 | 8, 9 | 916 | |||
XM_005253849.4 | XP_005253906.1 | 6, 7 | 595 | |||
XM_011537971.2 | XP_011536273.1 | 8, 9 | 871 | |||
XM_011537972.2 | XP_011536274.1 | 8, 9 | 744 | |||
XM_011537974.2 | XP_011536276.1 | 7, 8 | 747 | |||
XM_017018877.1 | XP_016874366.1 | 7, 8 | 660 | |||
XM_017018878.1 | XP_016874367.1 | 8, 9 | 864 | |||
XM_017018879.1 | XP_016874368.1 | 7, 8 | 800 | |||
XM_017018880.1 | XP_016874369.1 | 7, 8 | 708 | |||
XM_017018881.1 | XP_016874370.1 | 7, 8 | 800 | |||
XM_017018882.1 | XP_016874371.1 | 7, 8 | 652 | |||
GenBank mRNA | AK058176.1 | - | 4, 5 | 310 | ||
AK097665.1 | - | 9, 10 | 985 | |||
AK124109.1 | - | 8, 9 | 887 | |||
AK301826.1 | - | 8, 9 | 877 | |||
AK302512.1 | - | 6, 7 | 595 | |||
AY297459.1 | - | 8, 9 | 792 | |||
BC047645.1 | - | 9, 10 | 932 | 4828083 | ||
BC068031.1 | - | 8, 9 | 792 | 30378632 |