Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC38A6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172702.1 | NP_001166173.1 | 11 | 959 | ||
NM_153811.2 | NP_722518.2 | 11 | 959 | |||
NR_033344.1 | 11 | 1135 | ||||
XM_006720049.3 | XP_006720112.1 | 11 | 911 | |||
XM_006720050.1 | XP_006720113.1 | 7 | 402 | |||
XM_011536469.2 | XP_011534771.1 | 11 | 911 | |||
XM_017021020.1 | XP_016876509.1 | 11 | 911 | |||
XM_017021021.1 | XP_016876510.1 | 10 | 857 | |||
XM_017021022.1 | XP_016876511.1 | 11 | 1224 | |||
XM_017021023.1 | XP_016876512.1 | 11 | 911 | |||
XM_017021024.1 | XP_016876513.1 | 9 | 791 | |||
XM_017021025.1 | XP_016876514.1 | 11 | 1202 | |||
XM_017021026.1 | XP_016876515.1 | 11 | 1216 | |||
XM_017021027.1 | XP_016876516.1 | 9 | 580 | |||
XM_017021028.1 | XP_016876517.1 | 7 | 402 | |||
GenBank mRNA | AF070578.1 | - | 12 | 1047 | ||
AF543422.1 | - | 11 | 911 | |||
AK303839.1 | - | 11 | 938 | |||
BC110378.1 | - | 11 | 820 | 5737780 | ||
BX248072.1 | - | 11 | 780 | |||
BX648332.1 | - | 11 | 833 |