Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194287.2 | NP_919263.2 | 3 | 404 | ||
XM_011536479.1 | XP_011534781.1 | 3 | 772 | |||
XM_011536483.1 | XP_011534785.1 | 3 | 772 | |||
XM_011536487.1 | XP_011534789.1 | 3 | 772 | |||
XM_017021032.1 | XP_016876521.1 | 3 | 670 | |||
XM_017021033.1 | XP_016876522.1 | 3 | 772 | |||
XM_017021034.1 | XP_016876523.1 | 3 | 416 | |||
XM_017021035.1 | XP_016876524.1 | 3 | 567 | |||
XM_017021036.1 | XP_016876525.1 | 3 | 772 | |||
XM_017021037.1 | XP_016876526.1 | 3 | 1474 | |||
XM_017021038.1 | XP_016876527.1 | 3 | 308 | |||
XM_017021039.1 | XP_016876528.1 | 2 | 230 | |||
XM_017021040.1 | XP_016876529.1 | 3 | 229 | |||
GenBank mRNA | AK058106.1 | - | 3 | 414 | ||
AK125722.1 | - | 4 | 627 | |||
AK301762.1 | - | 3 | 550 | |||
BC062799.1 | - | 4 | 964 | |||
BC121159.2 | - | 3 | 252 | 40121413 | ||
BC121160.2 | - | 3 | 252 | 40121414 | ||
DC397584.1 | - | 3 | 555 |