Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC74A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194287.2 | NP_919263.2 | 3, 4 | 511 | ||
XM_011536479.1 | XP_011534781.1 | 3, 4 | 879 | |||
XM_011536483.1 | XP_011534785.1 | 3, 4 | 879 | |||
XM_011536487.1 | XP_011534789.1 | 3, 4 | 879 | |||
XM_017021032.1 | XP_016876521.1 | 3, 4 | 777 | |||
XM_017021033.1 | XP_016876522.1 | 3, 4 | 879 | |||
XM_017021034.1 | XP_016876523.1 | 3, 4 | 523 | |||
XM_017021035.1 | XP_016876524.1 | 3, 4 | 674 | |||
XM_017021036.1 | XP_016876525.1 | 3, 4 | 879 | |||
XM_017021037.1 | XP_016876526.1 | 3, 4 | 1581 | |||
XM_017021038.1 | XP_016876527.1 | 3, 4 | 415 | |||
XM_017021039.1 | XP_016876528.1 | 2, 3 | 337 | |||
XM_017021040.1 | XP_016876529.1 | 3, 4 | 336 | |||
GenBank mRNA | AK058106.1 | - | 3, 4 | 521 | ||
AK125722.1 | - | 4, 5 | 734 | |||
AK301762.1 | - | 3, 5 | 657 | |||
BC062799.1 | - | 4, 5 | 1071 | |||
BC121159.2 | - | 3, 4 | 359 | 40121413 | ||
BC121160.2 | - | 3, 4 | 359 | 40121414 |