Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284417.1 | NP_001271346.1 | 3 | 606 | ||
NM_001284418.1 | NP_001271347.1 | 3 | 607 | |||
NM_001284419.1 | NP_001271348.1 | 5 | 682 | |||
NM_001284420.1 | NP_001271349.1 | 2 | 316 | |||
NM_001284421.1 | NP_001271350.1 | 4 | 632 | |||
NM_001284422.1 | NP_001271351.1 | 5 | 732 | |||
NM_152449.3 | NP_689662.2 | 6 | 797 | |||
XM_006720397.3 | XP_006720460.2 | 2 | 330 | |||
XM_011521241.1 | XP_011519543.1 | 2 | 390 | |||
XM_011521242.2 | XP_011519544.1 | 2 | 908 | |||
XM_011521243.2 | XP_011519545.1 | 2 | 908 | |||
XM_011521244.1 | XP_011519546.1 | 3 | 959 | |||
XM_011521245.2 | XP_011519547.1 | 2 | 330 | |||
XM_017021926.1 | XP_016877415.1 | 4 | 823 | |||
XM_017021927.1 | XP_016877416.1 | 3 | 1144 | |||
XM_017021928.1 | XP_016877417.1 | 2 | 788 | |||
XM_017021929.1 | XP_016877418.1 | 4 | 985 | |||
XM_017021930.1 | XP_016877419.1 | 4 | 999 | |||
GenBank mRNA | AK057570.1 | - | 1 | 141 | ||
AK124852.1 | - | 4 | 935 | |||
AK290296.1 | - | 3 | 592 | |||
BC041097.1 | - | 6 | 774 | 5240362 | ||
BC084545.1 | - | 3 | 574 | 4590701 | ||
BG420645.1 | - | 3 | 562 | |||
CB989579.1 | - | 4 | 623 | |||
DA594971.1 | - | 2 | 290 | |||
HY131530.1 | - | 3 | 343 |