Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYSMD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284417.1 | NP_001271346.1 | 3 | 652 | ||
NM_001284418.1 | NP_001271347.1 | 3 | 653 | |||
NM_001284419.1 | NP_001271348.1 | 5 | 728 | |||
NM_001284420.1 | NP_001271349.1 | 2 | 362 | |||
NM_001284421.1 | NP_001271350.1 | 4 | 678 | |||
NM_001284422.1 | NP_001271351.1 | 5 | 778 | |||
NM_152449.3 | NP_689662.2 | 6 | 843 | |||
XM_006720397.3 | XP_006720460.2 | 2 | 376 | |||
XM_011521241.1 | XP_011519543.1 | 2 | 436 | |||
XM_011521242.2 | XP_011519544.1 | 2 | 954 | |||
XM_011521243.2 | XP_011519545.1 | 2 | 954 | |||
XM_011521244.1 | XP_011519546.1 | 3 | 1005 | |||
XM_011521245.2 | XP_011519547.1 | 2 | 376 | |||
XM_017021926.1 | XP_016877415.1 | 4 | 869 | |||
XM_017021927.1 | XP_016877416.1 | 3 | 1190 | |||
XM_017021928.1 | XP_016877417.1 | 2 | 834 | |||
XM_017021929.1 | XP_016877418.1 | 4 | 1031 | |||
XM_017021930.1 | XP_016877419.1 | 4 | 1045 | |||
GenBank mRNA | AK124852.1 | - | 4 | 981 | ||
AK290296.1 | - | 3 | 638 | |||
BC041097.1 | - | 6 | 820 | 5240362 | ||
CB989579.1 | - | 4 | 669 | |||
DA594971.1 | - | 2 | 336 |